COMH4211A9E5 Combi-Blocks, Inc.

[6-(4-Prop-2-ynylcarbamoyl-butyrylamino)-hexyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31381683
  • Cas Number : 1456806-44-2
  • Molecular Weight : 367.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0596
Availability details for
Quantity
  • Availability
  • Add to Cart