COMH4211A9E6 Combi-Blocks, Inc.

2-Methyl-2-(4-nitrophenyl)propan-1-amine

  • MDL number MFCD20485984
  • Cas Number : 180081-08-7
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0598
Availability details for
Quantity
  • Availability
  • Add to Cart