COMH4211A9EA Combi-Blocks, Inc.

3-Iodo-4-(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD11110815
  • Cas Number : 1131614-66-8
  • Molecular Weight : 346.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0604
Availability details for
Quantity
  • Availability
  • Add to Cart