COMH4211A9EB Combi-Blocks, Inc.

3-Ethoxy-2-iodopyridine

  • MDL number MFCD00234320
  • Cas Number : 111454-62-7
  • Molecular Weight : 249.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0605
Availability details for
Quantity
  • Availability
  • Add to Cart