COMH4211A9EC Combi-Blocks, Inc.

1-[2-Chloro-4-(2-methylpropoxy)phenyl]ethan-1-amine

  • MDL number MFCD20296975
  • Cas Number : 1343443-39-9
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0611
Availability details for
Quantity
  • Availability
  • Add to Cart