COMH4211A9EF Combi-Blocks, Inc.

5-Bromo-2-methoxy-n,n-dimethylbenzamide

  • MDL number MFCD13914856
  • Cas Number : 1030440-36-8
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0619
Availability details for
Quantity
  • Availability
  • Add to Cart