COMH4211A9F0 Combi-Blocks, Inc.

4-Chloro-2-(difluoromethyl)aniline

  • MDL number MFCD18392780
  • Cas Number : 1221424-65-2
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0623
Availability details for
Quantity
  • Availability
  • Add to Cart