COMH4211A9F1 Combi-Blocks, Inc.

2-[(4-Fluorophenyl)sulfanyl]aniline

  • MDL number MFCD11132342
  • Cas Number : 3169-85-5
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0626
Availability details for
Quantity
  • Availability
  • Add to Cart