COMH4211A9F2 Combi-Blocks, Inc.

N-(4-Bromobenzyl)-o-methylhydroxylamine

  • MDL number MFCD21100695
  • Cas Number : 916582-47-3
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0630

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0630
Availability details for
Quantity
  • Availability
  • Add to Cart