COMH4211A9F5 Combi-Blocks, Inc.

1-[(3,4-Difluorophenyl)methyl]-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD31579369
  • Cas Number : 2025179-14-8
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0634
Availability details for
Quantity
  • Availability
  • Add to Cart