COMH4211A9F6 Combi-Blocks, Inc.

1-(6-Methoxypyridin-3-yl)propan-1-one

  • MDL number MFCD18802862
  • Cas Number : 885229-42-5
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0638
Availability details for
Quantity
  • Availability
  • Add to Cart