COMH4211A9F9 Combi-Blocks, Inc.

4-(4-Acetyl-2-methoxy-5-nitrophenoxy)-butanoic acid ethyl ester

  • MDL number MFCD28143236
  • Cas Number : 1031702-80-3
  • Molecular Weight : 325.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0650
Availability details for
Quantity
  • Availability
  • Add to Cart