COMH4211A9FA Combi-Blocks, Inc.

1-(4-Bromo-2-isobutoxy-phenyl)-ethylamine

  • MDL number MFCD24341531
  • Cas Number : 1551635-62-1
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0654
Availability details for
Quantity
  • Availability
  • Add to Cart