COMH4211A9FB Combi-Blocks, Inc.

4-[(N-Methyl-aminooxy)-methyl]-benzoic acid methyl ester

  • MDL number MFCD31561571
  • Cas Number : 2270906-08-4
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0658
Availability details for
Quantity
  • Availability
  • Add to Cart