COMH4211A9FC Combi-Blocks, Inc.

Methyl 3-(((5-bromopyridin-2-yl)methoxy)carbonylamino)propanoate

  • MDL number MFCD31583357
  • Cas Number : 1404431-72-6
  • Molecular Weight : 317.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0661
Availability details for
Quantity
  • Availability
  • Add to Cart