COMH4211A9FD Combi-Blocks, Inc.

1H-Pyrazole-1-acetonitrile, alpha-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

  • MDL number MFCD29763680
  • Cas Number : 1612174-34-1
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propanenitrile
Inchi Code 1S/C12H18BN3O2/c1-9(6-14)16-8-10(7-15-16)13-17-11(2,3)12(4,5)18-13/h7-9H,1-5H3
Inchi Key CHZGNICJVVRUGL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0662
Availability details for
Quantity
  • Availability
  • Add to Cart