COMH4211AA02 Combi-Blocks, Inc.

(3-Chlorobenzylidene)propanedinitrile

  • MDL number MFCD00175209
  • Cas Number : 2972-73-8
  • Molecular Weight : 188.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-chlorobenzylidene)malononitrile
Inchi Code 1S/C10H5ClN2/c11-10-3-1-2-8(5-10)4-9(6-12)7-13/h1-5H
Inchi Key CGLGAIINXSHQBP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0674
Availability details for
Quantity
  • Availability
  • Add to Cart