COMH4211AA05 Combi-Blocks, Inc.

Benzaldehyde, 5-bromo-2-hydroxy-3-(phenylmethoxy)-

  • MDL number MFCD30572570
  • Cas Number : 697302-70-8
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0680
Availability details for
Quantity
  • Availability
  • Add to Cart