COMH4211AA0A Combi-Blocks, Inc.

1-(3-Hexyloxyphenyl)ethanone

  • MDL number MFCD00075025
  • Cas Number : 37062-71-8
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0690
Availability details for
Quantity
  • Availability
  • Add to Cart