COMH4211AA0B Combi-Blocks, Inc.

5-Bromo-1-(phenylacetyl)-2,3-dihydro-1h-indole

  • MDL number MFCD31583091
  • Cas Number : 928321-92-0
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0694
Availability details for
Quantity
  • Availability
  • Add to Cart