COMH4211AA0C Combi-Blocks, Inc.

2-Iodo-5-(phenylmethoxy)phenol

  • MDL number MFCD28128924
  • Cas Number : 90408-82-5
  • Molecular Weight : 326.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0695
Availability details for
Quantity
  • Availability
  • Add to Cart