COMH4211AA0E Combi-Blocks, Inc.

5-Bromo-2-chloro-n-(2,2,2-trifluoro-ethyl)-nicotinamide

  • MDL number MFCD16159778
  • Cas Number : 1247543-07-2
  • Molecular Weight : 317.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0700
Availability details for
Quantity
  • Availability
  • Add to Cart