COMH4211AA10 Combi-Blocks, Inc.

2-Amino-n-methoxy-n,3-dimethylbenzamide

  • MDL number MFCD12095378
  • Cas Number : 886574-89-6
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0706
Availability details for
Quantity
  • Availability
  • Add to Cart