COMH4211AA11 Combi-Blocks, Inc.

2-(4-Ethoxy-pyrazol-1-yl)-2-methyl-propionic acid

  • MDL number MFCD31579338
  • Cas Number : 2270911-24-3
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0711

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0711
Availability details for
Quantity
  • Availability
  • Add to Cart