COMH4211AA13 Combi-Blocks, Inc.

2-Methyl-3-[4-(propan-2-yloxy)phenyl]propanoic acid

  • MDL number MFCD23756138
  • Cas Number : 66735-07-7
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0717
Availability details for
Quantity
  • Availability
  • Add to Cart