COMH4211AA14 Combi-Blocks, Inc.

4-Aminobicyclo[2.2.2]octane-1-carboxamide

  • MDL number MFCD20702135
  • Cas Number : 863303-82-6
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0718
Availability details for
Quantity
  • Availability
  • Add to Cart