COMH4211AA15 Combi-Blocks, Inc.

3,6-Dimethylheptanoic acid

  • MDL number MFCD02258560
  • Cas Number : 44980-98-5
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0719

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0719
Availability details for
Quantity
  • Availability
  • Add to Cart