COMH4211AA16 Combi-Blocks, Inc.

5-Fluoro-1-(2-methoxy-ethyl)-1h-indole-2-carboxylic acid

  • MDL number MFCD31579376
  • Cas Number : 2270910-57-9
  • Molecular Weight : 237.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0720
Availability details for
Quantity
  • Availability
  • Add to Cart