COMH4211AA17 Combi-Blocks, Inc.

[1-(1-Phenylethyl)-1h-benzimidazol-2-yl]methanol

  • MDL number MFCD02630597
  • Cas Number : 380551-61-1
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0721
Availability details for
Quantity
  • Availability
  • Add to Cart