COMH4211AA18 Combi-Blocks, Inc.

Benzene, 1,1'-[(4-bromobutoxy)(1,1-dimethylethyl)silylene]bis-

  • MDL number MFCD31561476
  • Cas Number : 125010-58-4
  • Molecular Weight : 391.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0727
Availability details for
Quantity
  • Availability
  • Add to Cart