COMH4211AA19 Combi-Blocks, Inc.

tert-Butyl 3-formyl-4-nitro-1h-indole-1-carboxylate

  • MDL number MFCD26743646
  • Cas Number : 1195785-11-5
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0728
Availability details for
Quantity
  • Availability
  • Add to Cart