COMH4211AA1A Combi-Blocks, Inc.

3-Amino-5-methoxy-n,n-dimethylbenzamide

  • MDL number MFCD24046274
  • Cas Number : 1050513-84-2
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0729
Availability details for
Quantity
  • Availability
  • Add to Cart