COMH4211AA6B Combi-Blocks, Inc.

2-(1-(Isopropylsulfonyl)piperidin-4-yl)ethanol

  • MDL number MFCD27979888
  • Cas Number : 1435489-66-9
  • Molecular Weight : 235.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-(isopropylsulfonyl)piperidin-4-yl)ethan-1-ol
Inchi Code 1S/C10H21NO3S/c1-9(2)15(13,14)11-6-3-10(4-7-11)5-8-12/h9-10,12H,3-8H2,1-2H3
Inchi Key CUPNNHXOHAKMRV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0740
Availability details for
Quantity
  • Availability
  • Add to Cart