COMH4211AA6C Combi-Blocks, Inc.

3-Amino-5-ethoxy-n-methylbenzamide

  • MDL number MFCD24046269
  • Cas Number : 1500448-66-7
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0741
Availability details for
Quantity
  • Availability
  • Add to Cart