COMH4211AA6D Combi-Blocks, Inc.

1-[3-(2,2-Difluoro-propoxy)-phenyl]-ethylamine hydrochloride

  • MDL number MFCD31381726
  • Cas Number : 2204959-32-8
  • Molecular Weight : 251.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0743
Availability details for
Quantity
  • Availability
  • Add to Cart