COMH4211AA6E Combi-Blocks, Inc.

tert-Butyl 2-(4-bromo-2-(trifluoromethyl)phenoxy)ethylcarbamate

  • MDL number MFCD31557915
  • Cas Number : 1315465-28-1
  • Molecular Weight : 384.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0744
Availability details for
Quantity
  • Availability
  • Add to Cart