COMH4211AA6F Combi-Blocks, Inc.

2-Cyclopropylmethoxy-5-fluoro-benzylamine

  • MDL number MFCD11640273
  • Cas Number : 1096895-18-9
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0752
Availability details for
Quantity
  • Availability
  • Add to Cart