COMH4211AA70 Combi-Blocks, Inc.

2-([1-(4-Chlorophenyl)cyclobutyl]methoxy)acetic acid

  • MDL number MFCD31557926
  • Cas Number : 1356465-54-7
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0753
Availability details for
Quantity
  • Availability
  • Add to Cart