COMH4211AA71 Combi-Blocks, Inc.

3,5-Difluoro-4-methylbenzyl bromide

  • MDL number MFCD28790341
  • Cas Number : 1803833-70-6
  • Molecular Weight : 221.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0757
Availability details for
Quantity
  • Availability
  • Add to Cart