COMH4211AA72 Combi-Blocks, Inc.

3-(4-Aminomethyl-piperidin-1-yl)-2,2-dimethyl-propionic acid methyl ester hydrochloride

  • MDL number MFCD31557852
  • Cas Number : 2208788-27-4
  • Molecular Weight : 264.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0759
Availability details for
Quantity
  • Availability
  • Add to Cart