COMH4211AA74 Combi-Blocks, Inc.

3-Bromo-n-(cyclopropylmethyl)-4-iodobenzamide

  • MDL number MFCD13302258
  • Cas Number : 1147647-29-7
  • Molecular Weight : 380.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0764
Availability details for
Quantity
  • Availability
  • Add to Cart