COMH4211AA76 Combi-Blocks, Inc.

1-(1-(4-Trifluoromethylphenyl)ethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583078
  • Cas Number : 1604037-06-0
  • Molecular Weight : 366.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0776
Availability details for
Quantity
  • Availability
  • Add to Cart