COMH4211AA77 Combi-Blocks, Inc.

tert-Butyl 4-((5-bromopyridin-2-yl)oxy)piperidine-1-carboxylate

  • MDL number MFCD11877786
  • Cas Number : 194668-49-0
  • Molecular Weight : 357.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0779
Availability details for
Quantity
  • Availability
  • Add to Cart