COMH4211AA78 Combi-Blocks, Inc.

1-(Cyclopropylsulfonyl)piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD31613689
  • Cas Number : 2270907-10-1
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0781
Availability details for
Quantity
  • Availability
  • Add to Cart