COMH4211AA7A Combi-Blocks, Inc.

5-Bromo-2-methylsulfanyl-nicotinic acid tert-butyl ester

  • MDL number MFCD31557781
  • Cas Number : 2206265-58-7
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0787
Availability details for
Quantity
  • Availability
  • Add to Cart