COMH4211AA7B Combi-Blocks, Inc.

(2-Hydroxy-ethyl)-methyl-carbamic acid allyl ester

  • MDL number MFCD20287022
  • Cas Number : 1339652-25-3
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name allyl 2-hydroxyethyl(methyl)carbamate
Inchi Code 1S/C7H13NO3/c1-3-6-11-7(10)8(2)4-5-9/h3,9H,1,4-6H2,2H3
Inchi Key VIORRRMMYAZSRJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0789
Availability details for
Quantity
  • Availability
  • Add to Cart