COMH4211AA7C Combi-Blocks, Inc.

3-(3-Chlorophenoxy)-2,2-dimethylpropanoic acid

  • MDL number MFCD13874386
  • Cas Number : 946762-70-5
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0790
Availability details for
Quantity
  • Availability
  • Add to Cart