COMH4211AA7D Combi-Blocks, Inc.

5'-Methoxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-3-amine

  • MDL number MFCD31614082
  • Cas Number : 1639298-20-6
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0791
Availability details for
Quantity
  • Availability
  • Add to Cart