COMH4211AA7E Combi-Blocks, Inc.

2-(2-Cyclohexylmethoxy-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

  • MDL number MFCD31614095
  • Cas Number : 2246563-65-3
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0792
Availability details for
Quantity
  • Availability
  • Add to Cart