COMH4211AA7F Combi-Blocks, Inc.

3-[(Ethanesulfonyl)methyl]aniline

  • MDL number MFCD12171569
  • Cas Number : 1153557-35-7
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0793
Availability details for
Quantity
  • Availability
  • Add to Cart